10-Hydroxybenzo[h]quinoline 98%
Supplier: Apollo Scientific
Warning
OR315046-25G
APOSOR315046-25GEA 144 EUR
APOSOR315046-25G
10-Hydroxybenzo[h]quinoline 98%
10-Hydroxybenzo[h]quinoline
Formula: C₁₃H₉NO
MW: 195.22 g/mol
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00142350
CAS Number: 33155-90-7