1,4'-Bipiperidine-4'-carboxamide
Supplier: Apollo Scientific
OR315067-1G OR315067-250MG OR315067-500MG OR315067-5G
APOSOR315067-1GEA 184 EUR
APOSOR315067-1G APOSOR315067-250MG APOSOR315067-500MG APOSOR315067-5G
1,4'-Bipiperidine-4'-carboxamide
1,4'-Bipiperidine-4'-carboxamide
Formula: C₁₁H₂₁N₃O
MW: 211.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631148
CAS Number: 39633-82-4