5-Bromo-2-chloro-3-hydroxypyridine 98%
Supplier: Apollo Scientific
Synonyms: 5-Bromo-2-chloropyridin-3-ol
OR315101-100G OR315101-25G OR315101-5G
APOSOR315101-100GEA 1840 EUR
APOSOR315101-100G APOSOR315101-25G APOSOR315101-5G
5-Bromo-2-chloro-3-hydroxypyridine 98%
5-Bromo-2-chloro-3-hydroxypyridine
Formula: C₅H₃BrClNO
MW: 208.4400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08688592
CAS Number: 286946-77-8