2,5-Diformylfuran 98%
Supplier: Apollo Scientific
Synonyms: Furan-2,5-dicarbaldehyde, 2,5-Furandicarboxaldehyde
OR315125-1G OR315125-5G
APOSOR315125-1GEA 145 EUR
APOSOR315125-1G APOSOR315125-5G
2,5-Diformylfuran 98%
2,5-Diformylfuran
Formula: C₆H₄O₃
MW: 124.10 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Freezer
MDL Number: MFCD00671517
CAS Number: 823-82-5