9-Phenylcarbazole-2-boronic acid

Supplier: Apollo Scientific
Warning

Synonyms: (9-Phenyl-9H-carbazol-2-yl)boronic acid

OR315156-25G
APOSOR315156-25GEA 112 EUR
APOSOR315156-25G
9-Phenylcarbazole-2-boronic acid
9-Phenylcarbazole-2-boronic acid
Formula: C₁₈H₁₄BNO₂
MW: 287.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22207050
CAS Number: 1001911-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR