2-Iodophenyl isothiocyanate
Supplier: Apollo Scientific
Danger
Synonyms: 1-Iodo-2-isothiocyanatobenzene
OR315240-1G OR315240-25G OR315240-5G
APOSOR315240-1GEA 23.2 EUR
APOSOR315240-1G APOSOR315240-25G APOSOR315240-5G
2-Iodophenyl isothiocyanate
2-Iodophenyl isothiocyanate
Formula: C₇H₄INS
MW: 261.09 g/mol
Boiling Pt: 296
Melting Pt: 36…39 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00041057
CAS Number: 98041-44-2
UN: 2923
ADR: 8+6.1 III