(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl-p-toluenesulphonate 95%
Supplier: Apollo Scientific
Warning
Synonyms: (R)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methyl 4-methylbenzenesulphonate, D-(-)-1,2-Isopropylideneglycerol 3-(p-Toluenesulphonate)
OR315292-25G OR315292-5G
APOSOR315292-25GEA 227 EUR
APOSOR315292-25G APOSOR315292-5G
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl-p-toluenesulphonate 95%
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl-p-toluenesulphonate
Formula: C₁₃H₁₈O₅S
MW: 286.34 g/mol
Density: 1.208 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD00063233
CAS Number: 23788-74-1