Methyl-2-methyl-4-nitrobenzoate 98%
Supplier: Apollo Scientific
Synonyms: Methyl 2-methyl-4-nitrobenzoate
OR315322-100G
APOSOR315322-100GEA 108 EUR
APOSOR315322-100G
Methyl-2-methyl-4-nitrobenzoate 98%
Methyl 2-methyl-4-nitrobenzoate
Formula: C₉H₉NO₄
MW: 195.1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03618407
CAS Number: 62621-09-4