2-Bromo-9,10-anthraquinone

Supplier: Apollo Scientific
Warning

OR315395-100G
APOSOR315395-100GEA 201 EUR
APOSOR315395-100G
2-Bromo-9,10-anthraquinone
2-Bromo-9,10-anthraquinone
Formula: C₁₄H₇BrO₂
MW: 287.11 g/mol
Melting Pt: 208 °C
Storage Temperature: Ambient
MDL Number: MFCD00474541
CAS Number: 572-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR