3-Bromo-p-toluonitrile 98%
Supplier: Apollo Scientific
Warning
OR3157-25G OR3157-5G OR3157-100G
APOSOR3157-25GEA 24.8 EUR
APOSOR3157-25G APOSOR3157-5G APOSOR3157-100G
3-Bromo-p-toluonitrile 98%
3-Bromo-p-toluonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
Melting Pt: 41…45 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD06797818
CAS Number: 42872-74-2
UN: 3439
ADR: 6.1 III