2-Amino-4-bromophenol 98%

Supplier: Apollo Scientific
Danger

Synonyms: 5-Bromo-2-hydroxyaniline

OR3159-100G OR3159-25G
APOSOR3159-100GEA 112 EUR
APOSOR3159-100G APOSOR3159-25G
2-Amino-4-bromophenol 98%
2-Amino-4-bromophenol
Formula: C₆H₆BrNO
MW: 188.02 g/mol
Melting Pt: 130…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00235171
CAS Number: 40925-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR