5-Isopropyl-1,3-cyclohexanedione
Supplier: Apollo Scientific
Synonyms: 5-Isopropylcyclohexane-1,3-dione
OR316026-1G
APOSOR316026-1GEA 85.7 EUR
APOSOR316026-1G
5-Isopropyl-1,3-cyclohexanedione
5-Isopropyl-1,3-cyclohexanedione
Formula: C₉H₁₄O₂·H₂O
MW: 154.21 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00075263
CAS Number: 18456-87-6