5-Bromo-7H-pyrrolo[2,3-d]pyrimidine
Supplier: Apollo Scientific
Warning
OR317007-5G
APOSOR317007-5GEA 149 EUR
APOSOR317007-5G
5-Bromo-7H-pyrrolo[2,3-d]pyrimidine
5-Bromo-7H-pyrrolo[2,3-d]pyrimidine
Formula: C₆H₄BrN₃
MW: 198.0200 g/mol
Melting Pt: 199…201 °C
Storage Temperature: Ambient
MDL Number: MFCD01877574
CAS Number: 175791-49-8