5,8-Dioxaspiro[3.4]octane-2-methanol
Supplier: Apollo Scientific
Synonyms: 5,8-Dioxaspiro[3.4]octan-2-ylmethanol
OR317131-1G OR317131-5G
APOSOR317131-1GEA 180 EUR
APOSOR317131-1G APOSOR317131-5G
5,8-Dioxaspiro[3.4]octane-2-methanol
5,8-Dioxaspiro[3.4]octane-2-methanol
Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23106218
CAS Number: 545882-60-8