tert-Butyl-2,7-diazaspiro[3.5]nonane-7-carboxylate hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 2,7-diazaspiro[3.5]nonane-7-carboxylate hydrochloride
OR317165-1G OR317165-5G
APOSOR317165-1GEA 68.3 EUR
APOSOR317165-1G APOSOR317165-5G
tert-Butyl-2,7-diazaspiro[3.5]nonane-7-carboxylate hydrochloride
tert-Butyl-2,7-diazaspiro[3.5]nonane-7-carboxylate hydrochloride
Formula: C₁₂H₂₃ClN₂O₂
MW: 262.7762 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185086
CAS Number: 1023301-84-9