(R)-1-Boc-2-Azetidinemethanol
Supplier: Apollo Scientific
OR317182-1G OR317182-250MG
APOSOR317182-1GEA 209 EUR
APOSOR317182-1G APOSOR317182-250MG
(R)-1-Boc-2-Azetidinemethanol
(R)-1-Boc-2-Azetidinemethanol
Formula: C₉H₁₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD03790937
CAS Number: 161511-90-6