1,6-Diiodohexane 98%
Supplier: Apollo Scientific
Warning
OR3172-100G
APOSOR3172-100GEA 0 EUR
APOSOR3172-100G
1,6-Diiodohexane 98%
1,6-Diiodohexane
Formula: C₆H₁₂I₂
MW: 337.97 g/mol
Boiling Pt: 141…142 °C/10mmHg
Melting Pt: 44478 °C
Density: 2.052 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001103
CAS Number: 629-09-4
EINECS: 211-073-5