(R)-1-Boc-Azetidine-2-carboxylic acid methyl ester 97%
Supplier: Apollo Scientific
Warning
Synonyms: (R)-1-tert-Butyl 2-methyl azetidine-1,2-dicarboxylate
OR317245-1G OR317245-250MG
APOSOR317245-1GEA 78.3 EUR
APOSOR317245-1G APOSOR317245-250MG
(R)-1-Boc-Azetidine-2-carboxylic acid methyl ester 97%
(R)-1-Boc-Azetidine-2-carboxylic acid methyl ester
Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11042210
CAS Number: 1260593-39-2