3-Ethyl-3-oxetanemethanol 97%
Supplier: Apollo Scientific
Synonyms: (3-Ethyloxetan-3-yl)methanol, 2-Hydroxymethyl-2-ethyl-1,3-epoxypropane, 3-Hydroxymethyl-3-ethyloxetane
OR317261-100G OR317261-25G OR317261-500G
APOSOR317261-100GEA 24.8 EUR
APOSOR317261-100G APOSOR317261-25G APOSOR317261-500G
3-Ethyl-3-oxetanemethanol 97%
3-Ethyl-3-oxetanemethanol
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 96 °C/4mmHg
Density: 1.019 g/cm³ (25 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00190143
CAS Number: 3047-32-3
EINECS: 221-254-0