1-Iodobutane
Supplier: Apollo Scientific
Danger
Synonyms: 1-Butyl iodide
OR3174-100G OR3174-500G
APOSOR3174-100GEA 72.5 EUR
APOSOR3174-100G APOSOR3174-500G
1-Iodobutane
1-Iodobutane
Formula: C₄H₉I
MW: 184.02 g/mol
Boiling Pt: 130…131 °C
Melting Pt: –103 °C
Density: 1.617 g/cm³ (25 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00001098
CAS Number: 542-69-8
EINECS: 208-824-4
UN: 1993
ADR: 3,III