5-Methanesulphonyl-1H-indole-2-carboxylic acid
Supplier: Apollo Scientific
Warning
OR318001-1G OR318001-5G OR318001-10G
APOSOR318001-1GEA 227 EUR
APOSOR318001-1G APOSOR318001-5G APOSOR318001-10G
5-Methanesulphonyl-1H-indole-2-carboxylic acid
5-Methanesulphonyl-1H-indole-2-carboxylic acid
Formula: C₁₀H₉NO₄S
MW: 239.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10688267
CAS Number: 367501-41-5