[R,S]-tert-Butyl-3-amino-2-methylpropionate 95+%
Supplier: Apollo Scientific
OR318014-1G OR318014-10G OR318014-5G
APOSOR318014-1GEA 578 EUR
APOSOR318014-1G APOSOR318014-10G APOSOR318014-5G
[R,S]-tert-Butyl-3-amino-2-methylpropionate 95+%
[R,S]-tert-Butyl-3-amino-2-methylpropionate
Formula: C₈H₁₇NO₂
MW: 159.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10688272
CAS Number: 120378-12-3