3-(5-Imidazolyl)pyridine 95+%
Supplier: Apollo Scientific
Warning
OR318018-1G OR318018-5G OR318018-10G
APOSOR318018-1GEA 578 EUR
APOSOR318018-1G APOSOR318018-5G APOSOR318018-10G
3-(5-Imidazolyl)pyridine 95+%
3-(5-Imidazolyl)pyridine
Formula: C₈H₇N₃
MW: 145.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10565593
CAS Number: 51746-85-1