1-Ethyl-3-methyl-4-nitro-1H-pyrazole-5-carboxylic acid 95+%
Supplier: Apollo Scientific
OR318019-1G OR318019-10G OR318019-5G
APOSOR318019-1GEA 200 EUR
APOSOR318019-1G APOSOR318019-10G APOSOR318019-5G
1-Ethyl-3-methyl-4-nitro-1H-pyrazole-5-carboxylic acid 95+%
1-Ethyl-3-methyl-4-nitro-1H-pyrazole-5-carboxylic acid
Formula: C₇H₉N₃O₄
Storage Temperature: Ambient
MDL Number: MFCD00207991
CAS Number: 26308-39-4