5-Hydroxy-2-methoxybenzaldehyde 95+%
Supplier: Apollo Scientific
Warning
OR318031-1G OR318031-5G
APOSOR318031-1GEA 71.6 EUR
APOSOR318031-1G APOSOR318031-5G
5-Hydroxy-2-methoxybenzaldehyde 95+%
5-Hydroxy-2-methoxybenzaldehyde
Formula: C₈H₈O₃
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD03820046
CAS Number: 35431-26-6