5-Bromo-8-chloro-1,7-naphthyridine 90%
Supplier: Apollo Scientific
OR318076-1G OR318076-250MG
APOSOR318076-1GEA 232 EUR
APOSOR318076-1G APOSOR318076-250MG
5-Bromo-8-chloro-1,7-naphthyridine 90%
5-Bromo-8-chloro-1,7-naphthyridine
Formula: C₈H₄BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD08567946
CAS Number: 909649-06-5