[4-(Methylsulphonyl)-2-nitrophenyl]methanol 90%
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Methylsulphonyl)-2-nitrobenzyl alcohol
OR318084-1G OR318084-10G OR318084-5G
APOSOR318084-1GEA 294 EUR
APOSOR318084-1G APOSOR318084-10G APOSOR318084-5G
[4-(Methylsulphonyl)-2-nitrophenyl]methanol 90%
[4-(Methylsulphonyl)-2-nitrophenyl]methanol
Formula: C₈H₉NO₅S
MW: 231.23 g/mol
Melting Pt: 134…137 °C
Storage Temperature: Ambient
MDL Number: MFCD06203102
CAS Number: 773873-23-7