trans-1,4-Dibromo-2-butene
Supplier: Apollo Scientific
Danger
Synonyms: (E)-1,4-Dibromobut-2-ene, (E)-1,4-Dibromo-2-butene
OR3193-100G OR3193-25G OR3193-500G
APOSOR3193-100GEA 36.4 EUR
APOSOR3193-100G APOSOR3193-25G APOSOR3193-500G
trans-1,4-Dibromo-2-butene
trans-1,4-Dibromo-2-butene
Formula: C₄H₆Br₂
Boiling Pt: 205 °C
Melting Pt: 48…53 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000249
CAS Number: 821-06-7
EINECS: 212-472-7
UN: 2923
ADR: 8+6.1,II