trans-1,2-Cyclohexanedimethanol
Supplier: Apollo Scientific
Warning
Synonyms: trans-Cyclohexane-1,2-diyldimethanol, trans-1,2-Bis(hydroxymethyl)cyclohexane
OR3201-1G OR3201-5G
APOSOR3201-1GEA 40.6 EUR
APOSOR3201-1G APOSOR3201-5G
trans-1,2-Cyclohexanedimethanol
trans-1,2-Cyclohexanedimethanol
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 113 °C/ 0.03 kPa
Melting Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD09260493
CAS Number: 25712-33-8