1-[4-(Methylsulphanyl)phenyl]-1,4-pentanedione 95%
Supplier: Apollo Scientific
OR32086-1G OR32086-5G
APOSOR32086-1GEA 270 EUR
APOSOR32086-1G APOSOR32086-5G
1-[4-(Methylsulphanyl)phenyl]-1,4-pentanedione 95%
1-[4-(Methylsulphanyl)phenyl]-1,4-pentanedione
Formula: C₁₂H₁₄O₂S
MW: 222.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08056601
CAS Number: 189501-33-5