1-Methyl-1,5,6,7-tetrahydroindazol-4-one 95%
Supplier: Apollo Scientific
Synonyms: 1-Methyl-6,7-dihydro-1H-indazol-4(5H)-one
OR32091-1G OR32091-5G
APOSOR32091-1GEA 339 EUR
APOSOR32091-1G APOSOR32091-5G
1-Methyl-1,5,6,7-tetrahydroindazol-4-one 95%
1-Methyl-1,5,6,7-tetrahydroindazol-4-one
Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00930489
CAS Number: 85302-16-5