Methyl-3-bromo-4-iodobenzoate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3-bromo-4-iodobenzoate
OR321001-1G OR321001-5G
APOSOR321001-1GEA 21.9 EUR
APOSOR321001-1G APOSOR321001-5G
Methyl-3-bromo-4-iodobenzoate
Methyl 3-bromo-4-iodobenzoate
Formula: C₈H₆BrIO₂
MW: 340.9400 g/mol
Melting Pt: 56…60 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07778512
CAS Number: 249647-24-3