tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Boc-DL-Prolinol, tert-Butyl 2-(hydroxymethyl)pyrrolidine-1-carboxylate
OR321308-5G OR321308-25G
APOSOR321308-5GEA 58 EUR
APOSOR321308-5G APOSOR321308-25G
tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate 97%
tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate
Formula: C₁₀H₁₉NO₃
MW: 201.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01456556
CAS Number: 170491-63-1