2-Bromo-3-methylpyrazine 97%
Supplier: Apollo Scientific
Warning
OR321322-1G OR321322-5G
APOSOR321322-1GEA 49.3 EUR
APOSOR321322-1G APOSOR321322-5G
2-Bromo-3-methylpyrazine 97%
2-Bromo-3-methylpyrazine
Formula: C₅H₅BrN₂
MW: 173.0100 g/mol
Boiling Pt: 194.8 °C/760mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD08705769
CAS Number: 120984-76-1