1H-Pyrrolo[3,2-b]pyridin-5-amine hydrochloride 97%
Supplier: Apollo Scientific
Warning
OR321325-1G OR321325-100MG OR321325-250MG
APOSOR321325-1GEA 214 EUR
APOSOR321325-1G APOSOR321325-100MG APOSOR321325-250MG
1H-Pyrrolo[3,2-b]pyridin-5-amine hydrochloride 97%
1H-Pyrrolo[3,2-b]pyridin-5-amine hydrochloride
Formula: C₇H₈ClN₃
MW: 169.6100 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16875696
CAS Number: 1260771-53-6