Ethyl 1-Boc-5-oxoazepane-4-carboxylate 97%
Supplier: Apollo Scientific
Synonyms: 1-tert-Butyl 4-ethyl 5-oxoazepane-1,4-dicarboxylate
OR321470-1G OR321470-5G
APOSOR321470-1GEA 119 EUR
APOSOR321470-1G APOSOR321470-5G
Ethyl 1-Boc-5-oxoazepane-4-carboxylate 97%
Ethyl-1-boc-5-oxoazepane-4-carboxylate
Formula: C₁₄H₂₃NO₅
MW: 285.336 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788451
CAS Number: 141642-82-2