Methyl-2-(3-piperidinyl)benzoate hydrochloride
Supplier: Apollo Scientific
OR321530-1G OR321530-250MG
APOSOR321530-1GEA 702 EUR
APOSOR321530-1G APOSOR321530-250MG
Methyl-2-(3-piperidinyl)benzoate hydrochloride
Methyl-2-(3-piperidinyl)benzoate hydrochloride
Formula: C₁₃H₁₈ClNO₂
MW: 255.75 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08751527
CAS Number: 1203682-56-7