N-Allyl-3-chlorobenzenecarboxamide 95%

Supplier: Apollo Scientific

Synonyms: 3-Chloro-N-(prop-2-en-1-yl)benzamide

OR32200-1G
APOSOR32200-1GEA 180 EUR
APOSOR32200-1G
N-Allyl-3-chlorobenzenecarboxamide 95%
N-Allyl-3-chlorobenzenecarboxamide
Formula: C₁₀H₁₀ClNO
MW: 195.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01181345
CAS Number: 35306-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR