Ethyl chloroacetate 99%
Supplier: Apollo Scientific
OR322004-500G OR322004-2.5KG
APOSOR322004-500GEA 87 EUR
APOSOR322004-500G APOSOR322004-2.5KG
Ethyl chloroacetate 99%
Ethyl chloroacetate
Formula: C₄H₇ClO₂
Boiling Pt: 143 °C
Melting Pt: –26 °C
Density: 1.145 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000932
CAS Number: 105-39-5
UN: 1181
ADR: 6.1+3,II