trans-2,3-Epoxybutane
Supplier: Apollo Scientific
OR322036-1G OR322036-5G OR322036-25G
APOSOR322036-1GEA 72.5 EUR
APOSOR322036-1G APOSOR322036-5G APOSOR322036-25G
trans-2,3-Epoxybutane
trans-2,3-Epoxybutane
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 54…55 °C
Density: 0.804 g/cm³ (25 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD00005129
CAS Number: 21490-63-1
UN: 3271
ADR: 3,II