1-Aminomethyl-1-cyclohexanol hydrochloride 97%
Supplier: Apollo Scientific
OR322146-1G OR322146-5G
APOSOR322146-1GEA 132 EUR
APOSOR322146-1G APOSOR322146-5G
1-Aminomethyl-1-cyclohexanol hydrochloride 97%
1-Aminomethyl-1-cyclohexanol hydrochloride
Formula: C₇H₁₆ClNO
Melting Pt: 217…219 °C
Storage Temperature: Ambient
CAS Number: 19968-85-5