2-Chloroethyl isothiocyanate 97%
Supplier: Apollo Scientific
Danger
OR322152-1G OR322152-5G
APOSOR322152-1GEA 68.3 EUR
APOSOR322152-1G APOSOR322152-5G
2-Chloroethyl isothiocyanate 97%
2-Chloroethyl isothiocyanate
Formula: C₃H₄ClNS
MW: 121.59 g/mol
Boiling Pt: 79…80 °C/13mmHg
Density: 1.265 g/cm³ (25 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00041125
CAS Number: 6099-88-3
UN: 3286
ADR: 3+6.1+8 II