5-Chloro-2-nitro-p-toluidine 95%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Chloro-4-methyl-2-nitroaniline
OR322344-25G
APOSOR322344-25GEA 195 EUR
APOSOR322344-25G
5-Chloro-2-nitro-p-toluidine 95%
5-Chloro-2-nitro-p-toluidine
Formula: C₇H₇ClN₂O₂
MW: 186.60 g/mol
Melting Pt: 159…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00034066
CAS Number: 7149-80-6