Methyl acrylate 99%
Supplier: Apollo Scientific
Danger
Synonyms: Acrylic acid methyl ester
OR322452-500ML OR322452-2500M
APOSOR322452-500MLEA 72.5 EUR
APOSOR322452-500ML APOSOR322452-2500M
Methyl acrylate 99%
Methyl acrylate (monomer)
Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 80 °C
Melting Pt: –75 °C
Density: 0.956 g/cm³ (25 °C)
Flash Pt: –3 °C
Storage Temperature: Ambient
MDL Number: MFCD00008627
CAS Number: 96-33-3
UN: 1919
ADR: 3,II