2'-Methoxy-[1,1'-biphenyl]-3-amine
Supplier: Apollo Scientific
Synonyms: 2'-Methoxybiphenyl-3-ylamine, 3-(2-Methoxyphenyl)aniline
OR322524-1G OR322524-500MG
APOSOR322524-1GEA 328 EUR
APOSOR322524-1G APOSOR322524-500MG
2'-Methoxy-[1,1'-biphenyl]-3-amine
2'-Methoxy-[1,1'-biphenyl]-3-amine
Formula: C₁₃H₁₃NO
MW: 199.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424666
CAS Number: 96923-01-2