3-Acetylamino-4-methoxybenzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
OR322585-1G OR322585-250MG
APOSOR322585-1GEA 151 EUR
APOSOR322585-1G APOSOR322585-250MG
3-Acetylamino-4-methoxybenzenesulphonyl chloride
3-Acetylamino-4-methoxybenzenesulphonyl chloride
Formula: C₉H₁₀ClNO₄S
MW: 263.7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00129812
CAS Number: 3746-67-6
UN: 1759
ADR: 8,III