Ethyl-(S)-(+)-2-pyrrolidinone-5-carboxylate 98%
Supplier: Apollo Scientific
Synonyms: Ethyl (S)-5-oxopyrrolidine-2-carboxylate, H-Pyr-OEt, Ethyl (S)-(+)-2-pyrrolidone-5-carboxylate
OR322668-500G
APOSOR322668-500GEA 112 EUR
APOSOR322668-500G
Ethyl-(S)-(+)-2-pyrrolidinone-5-carboxylate 98%
Ethyl-(S)-(+)-2-pyrrolidinone-5-carboxylate
Formula: C₇H₁₁NO₃
MW: 157.169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064497
CAS Number: 7149-65-7