4-Chloro-1-methyl[1,2,4]triazolo[4,3-a]quinoxaline 95%
Supplier: Apollo Scientific
OR32297-1G OR32297-5G
APOSOR32297-1GEA 336 EUR
APOSOR32297-1G APOSOR32297-5G
4-Chloro-1-methyl[1,2,4]triazolo[4,3-a]quinoxaline 95%
4-Chloro-1-methyl[1,2,4]triazolo[4,3-a]quinoxaline
Formula: C₁₀H₇ClN₄
MW: 218.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00140733
CAS Number: 91895-39-5