2-(Trimethylsilyl)ethoxymethyl chloride 90%
Supplier: Apollo Scientific
Danger
Synonyms: 2-(Chloromethoxy)ethyltrimethylsilane, 2-(Trimethylsilyl)ethoxymethyl Chloride, SEM-chloride
OR323097-100G OR323097-25G OR323097-10G
APOSOR323097-100GEA 393 EUR
APOSOR323097-100G APOSOR323097-25G APOSOR323097-10G
2-(Trimethylsilyl)ethoxymethyl chloride 90%
2-(Trimethylsilyl)ethoxymethyl chloride
Formula: C₆H₁₅ClOSi
MW: 166.72 g/mol
Boiling Pt: 170…172 °C
Flash Pt: 42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009919
CAS Number: 76513-69-4
UN: 2920
ADR: 8+3,II