(R)-(+)-2-Methyl-2-propanesulphinamide 98%
Supplier: Apollo Scientific
Warning
Synonyms: (R)-2-methylpropane-2-sulphinamide, R)-(+)-tert-Butyl sulphinamide, (R)-2-Methyl-2-propanesulphinamide, (R)-(+)-tert-Butylsulphinamide, (R)-(+)-t-Butylsulphinamide
OR3233-100G OR3233-25G OR3233-500G
APOSOR3233-100GEA 84.1 EUR
APOSOR3233-100G APOSOR3233-25G APOSOR3233-500G
(R)-(+)-2-Methyl-2-propanesulphinamide 98%
(R)-(+)-2-Methyl-2-propanesulphinamide
Formula: C₄H₁₁NOS
MW: 121.20 g/mol
Melting Pt: 103…107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05861479
CAS Number: 196929-78-9